Assay Details
Target Gene Details
Entrez Gene ID: | 226982 |
Gene Name: | eukaryotic translation initiation factor 5B |
Gene Aliases: |
A030003E17Rik, BC018347, IF2 |
Location: |
Chr.1:37998000-38055579 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5b | NM_198303.2 | 24 | 6137 | NP_938045.2 |
| AK136813.1 | 1 | 839 | ||
| AK173016.1 | 8 | 3210 | BAD32294.1 |
Target Gene Details
Entrez Gene ID: | 56210 |
Gene Name: | REV1 homolog (S. cerevisiae) |
Gene Aliases: |
1110027I23Rik, AU022044, Rev1l |
Location: |
Chr.1:38052786-38129919 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rev1 | NM_019570.3 | 21 | 3667 | NP_062516.2 |
| AB057418.1 | 20 | 3456 | BAB64933.1 | |
| AF179302.1 | 20 | 3456 | AAF23323.1 | |
| BC058093.1 | 21 | 3667 | AAH58093.1 |
More Information
Set Membership: |
|

Genomic Map