Assay Details
Target Gene Details
Entrez Gene ID: | 104009 |
Gene Name: | quiescin Q6 sulfhydryl oxidase 1 |
Gene Aliases: |
1300003H02Rik, QSOX, Qscn6, SOx |
Location: |
Chr.1:155778155-155812899 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Qsox1 | NM_001024945.1 | NP_001020116.1 | ||
| NM_023268.2 | NP_075757.1 | |||
| AB044284.1 | BAB21936.1 | |||
| AK004880.1 | BAB23638.1 | |||
| AK083938.1 | BAC39073.1 | |||
| AK166839.1 | BAE39061.1 | |||
| AK169877.1 | BAE41429.1 | |||
| AK169920.1 | BAE41459.1 | |||
| AK170449.1 | BAE41807.1 | |||
| BC034131.1 | AAH34131.1 | |||
| BC076590.1 | AAH76590.1 |
More Information
Set Membership: |
|

Genomic Map