Assay Details
Target Gene Details
Entrez Gene ID: | 240816 |
Gene Name: | regulator of G-protein signaling like 1 |
Gene Aliases: |
4930415K13Rik, Gm202, Rgsl2 |
Location: |
Chr.1:153777929-153847342 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgsl1 | NM_001243223.1 | NP_001230152.1 | ||
| AK029630.1 | BAC26537.1 | |||
| AK161463.1 |

Genomic Map