Assay Details
Target Gene Details
Entrez Gene ID: | 19821 |
Gene Name: | ring finger protein 2 |
Gene Aliases: |
AI326319, AI450156, AU019207, Ring1B, dinG |
Location: |
Chr.1:151469406-151500923 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf2 | NM_011277.2 | NP_035407.1 | ||
| AK012358.2 | BAB28186.1 | |||
| AK013124.1 | BAB28663.1 | |||
| AK030319.1 | BAC26898.1 | |||
| AK090066.1 | BAC41075.1 | |||
| AK145767.1 | BAE26638.1 | |||
| BC020122.1 | AAH20122.1 | |||
| CT010297.1 | CAJ18505.1 | |||
| Y12880.1 | CAA73380.1 |
More Information
Set Membership: |
|

Genomic Map