Assay Details
Target Gene Details
Entrez Gene ID: | 67223 |
Gene Name: | ribosomal RNA processing 15 homolog (S. cerevisiae) |
Gene Aliases: |
2810430M08Rik, 5430437H21Rik |
Location: |
Chr.1:186721086-186749358 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rrp15 | NM_026041.2 | NP_080317.3 | ||
| AK010658.1 | BAB27096.1 | |||
| AK013204.1 | ||||
| AK013217.1 | BAB28719.2 | |||
| AK087997.1 | BAC40083.1 | |||
| AK169088.1 | BAE40872.1 | |||
| BC006787.1 | AAH06787.1 |

Genomic Map