Assay Details
Target Gene Details
Entrez Gene ID: | 20724 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 5 |
Gene Aliases: |
1110036M19Rik, AI462524, AI646751, Maspin, PI-5, Spi7, ovalbumin |
Location: |
Chr.1:106861117-106883350 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb5 | NM_009257.3 | NP_033283.1 | ||
| AK136532.1 | BAE23032.1 | |||
| AK145220.1 | BAE26308.1 | |||
| BC005434.1 | AAH05434.1 | |||
| U54705.1 | AAB06042.1 |
More Information
Set Membership: |
|

Genomic Map