Assay Details
Target Gene Details
Entrez Gene ID: | 76816 |
Gene Name: | serologically defined colon cancer antigen 8 |
Gene Aliases: |
2700048G21Rik, 5730470G24Rik, Cccap |
Location: |
Chr.1:176814526-177020437 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdccag8 | NM_029756.3 | 11 | 1629 | NP_084032.1 |
| AK012392.1 | 11 | 1417 | BAB28209.1 | |
| AK051683.1 | 12 | 1640 | BAC34719.1 | |
| AK132316.1 | 11 | 1425 | BAE21100.1 | |
| AK133622.1 | 11 | 1373 | BAE21753.1 | |
| AK136921.1 | 12 | 1660 | BAE23170.1 | |
| BC016244.1 | 12 | 1524 |

Genomic Map