Assay Details
Target Gene Details
Entrez Gene ID: | 20344 |
Gene Name: | selectin, platelet |
Gene Aliases: |
CD62P, GMP-140, Grmp, LECAM3, PADGEM |
Location: |
Chr.1:164115264-164150026 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Selp | NM_011347.2 | 6 | 952 | NP_035477.1 |
| BC109158.1 | 5 | 838 | AAI09159.1 | |
| BC109159.1 | 5 | 838 | AAI09160.1 | |
| M72332.1 | 6 | 952 | AAA37712.1 | |
| M87861.1 | 5 | 838 | AAA40008.1 |

Genomic Map