Assay Details
Target Gene Details
Entrez Gene ID: | 98366 |
Gene Name: | small ArfGAP 1 |
Gene Aliases: |
1700056O10Rik, 4921514B13Rik, 4921525H11Rik, AI462175 |
Location: |
Chr.1:23844825-23922450 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smap1 | NM_001290683.1 | 7 | 611 | NP_001277612.1 |
| NM_028534.3 | 7 | 881 | NP_082810.1 | |
| NR_110972.1 | 7 | 607 | ||
| AK014888.1 | 7 | 567 | ||
| AK014966.1 | 7 | 565 | ||
| AK132868.1 | 7 | 608 | ||
| BC006946.1 | 7 | 789 | AAH06946.1 |

Genomic Map