Assay Details
Target Gene Details
Entrez Gene ID: | 641376 |
Gene Name: | translocase of outer mitochondrial membrane 40 homolog-like (yeast) |
Gene Aliases: |
- |
Location: |
Chr.1:171217802-171222514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tomm40l | NM_001037170.2 | NP_001032247.1 | ||
| AK012264.1 | BAB28127.1 | |||
| AK028388.1 | BAC25924.1 | |||
| AK029485.1 | BAC26471.1 | |||
| AK177205.1 | ||||
| AK186544.1 | ||||
| BC058978.1 | AAH58978.1 |
More Information
Set Membership: |
|

Genomic Map