Assay Details
Target Gene Details
Entrez Gene ID: | 240832 |
Gene Name: | torsin A interacting protein 2 |
Gene Aliases: |
1110020D10Rik, 15kDa, A130072J07, AA103493, AW060462, AW610675, C77739, Ifrg15, Lull1 |
Location: |
Chr.1:156035664-156068861 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip2 | NM_172843.4 | 4 | 915 | NP_766431.3 |
| AK038024.1 | 4 | 1177 | BAC29923.1 | |
| AK080959.1 | 2 | 194 | BAC38096.1 | |
| BC066790.1 | 2 | 121 | AAH66790.1 | |
| CF164425.1 |

Genomic Map