Assay Details
Target Gene Details
Entrez Gene ID: | 14897 |
Gene Name: | thyroid hormone receptor interactor 12 |
Gene Aliases: |
1110036I07Rik, 6720416K24Rik, AA410158, Gtl6, TRIP-12 |
Location: |
Chr.1:84721189-84850008 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip12 | NM_133975.4 | NP_598736.4 | ||
| AK033327.1 | ||||
| AW543112.1 | ||||
| BC034113.1 | ||||
| CF747879.1 |
More Information
Set Membership: |
|

Genomic Map