Assay Details
Target Gene Details
Entrez Gene ID: | 20822 |
Gene Name: | TROVE domain family, member 2 |
Gene Aliases: |
1810007I17Rik, A530054J02Rik, AI646302, SS-A/Ro, Ssa, Ssa2 |
Location: |
Chr.1:143750790-143777051 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trove2 | NM_013835.2 | NP_038863.1 | ||
| AK076167.1 | ||||
| AK167379.1 | BAE39472.1 | |||
| AK167555.1 | BAE39620.1 | |||
| AK167645.1 | BAE39696.1 | |||
| BC051974.1 | AAH51974.1 | |||
| BC052380.1 | ||||
| L81154.1 | AAL77518.1 | |||
| U66843.1 | AAC53142.1 |
More Information
Set Membership: |
|

Genomic Map