Assay Details
Target Gene Details
Entrez Gene ID: | 209456 |
Gene Name: | transformation related protein 53 binding protein 2 |
Gene Aliases: |
53BP2, AI746547, ASPP2, PPP1R13A, Tp53bp2, X98550 |
Location: |
Chr.1:182409167-182462436 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trp53bp2 | NM_173378.2 | NP_775554.2 | ||
| AK134556.1 | BAE22185.1 | |||
| BC030894.1 | AAH30894.1 |
More Information
Set Membership: |
|

Genomic Map