Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igfn1 |
Target Gene Details
Entrez Gene ID: | 66241 |
Gene Name: | transmembrane protein 9 |
Gene Aliases: |
1500015G18Rik, AW545782 |
Location: |
Chr.1:136008219-136035030 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem9 | NM_001160145.1 | NP_001153617.1 | ||
| NM_001160146.1 | NP_001153618.1 | |||
| NM_025439.3 | NP_079715.1 | |||
| AK005248.1 | BAB23903.1 | |||
| AK021225.1 | BAB32335.1 | |||
| AK134909.1 | BAE22335.1 | |||
| AK154423.1 | BAE32575.1 | |||
| AK190183.1 | ||||
| BC016524.1 | AAH16524.1 | |||
| BQ918002.1 | ||||
| CA460035.1 | ||||
| CX208138.1 |

Genomic Map