Assay Details
Target Gene Details
Entrez Gene ID: | 21934 |
Gene Name: | tumor necrosis factor receptor superfamily, member 11a, NFKB activator |
Gene Aliases: |
Ly109, ODFR, OFE, Rank, TRANCE-R, mRANK |
Location: |
Chr.1:105780723-105847981 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf11a | NM_009399.3 | 3 | 250 | NP_033425.3 |
| AF019046.1 | 3 | 245 | AAB86810.1 | |
| AK020374.1 | 3 | 223 | ||
| AK138990.1 | 3 | 256 |
More Information
Set Membership: |
|

Genomic Map