Assay Details
Target Gene Details
Entrez Gene ID: | 66637 |
Gene Name: | tRNA splicing endonuclease 15 homolog (S. cerevisiae) |
Gene Aliases: |
5730449L18Rik, AL023077, Sen15 |
Location: |
Chr.1:152370735-152386682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsen15 | NM_025677.3 | NP_079953.2 | ||
| AK004261.1 | ||||
| AK011608.1 | ||||
| AK017650.1 | BAB30855.1 | |||
| AK169429.1 | ||||
| BC024342.1 | AAH24342.1 | |||
| BC145701.1 |

Genomic Map