Assay Details
Target Gene Details
Entrez Gene ID: | 71872 |
Gene Name: | aldehyde oxidase 4 |
Gene Aliases: |
2310003G12Rik, Aoh2 |
Location: |
Chr.1:58210397-58268600 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aox4 | NM_023631.2 | 1 | 122 | NP_076120.2 |
| AF233581.1 | 1 | 49 | AAF98385.1 | |
| AK081746.1 | 1 | 68 | ||
| AK158427.1 | 1 | 123 | BAE34503.1 | |
| AK160953.1 | 1 | 88 | ||
| BC117975.1 | 1 | 49 | ||
| BC117976.1 | 1 | 49 |

Genomic Map