Assay Details
Target Gene Details
Entrez Gene ID: | 226970 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 4 |
Gene Aliases: |
9330140K16Rik, Asef, ENSMUSG00000070955 |
Location: |
Chr.1:34801722-34812754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgef4 | NM_183019.2 | NP_898840.2 | ||
| AK031310.1 | 5 | 871 | ||
| AK034013.1 | ||||
| AK138598.1 | ||||
| AK163424.1 | BAE37343.1 | |||
| AK169449.1 | ||||
| AK169957.1 | BAE41480.1 | |||
| BC139371.1 | ||||
| BC139397.1 |
More Information
Set Membership: |
|

Genomic Map