Assay Details
Target Gene Details
Entrez Gene ID: | 22596 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 5 |
Gene Aliases: |
AI314015, Ku80, Ku86 |
Location: |
Chr.1:72307417-72394953 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc5 | NM_009533.2 | 2 | 207 | NP_033559.2 |
| AF166486.1 | 2 | 102 | AAD49720.1 | |
| AK010416.1 | 2 | 96 | BAB26920.3 | |
| AK081633.1 | 2 | 208 | BAC38276.1 | |
| AK089944.1 | 2 | 204 | ||
| AK165470.1 | 2 | 175 | BAE38207.1 | |
| AK167312.1 | 2 | 96 | BAE39415.1 | |
| AK168913.1 | 2 | 96 | BAE40726.1 | |
| AK169838.1 | 2 | 96 | BAE41402.1 | |
| AK182445.1 | 1 | 68 | ||
| AK200491.1 | ||||
| BC029218.1 | 2 | 79 | AAH29218.1 | |
| BC051660.1 | 2 | 147 | AAH51660.1 | |
| X66323.1 | 2 | 108 | CAA46999.1 |
More Information
Set Membership: |
|

Genomic Map