Assay Details
Target Gene Details
Entrez Gene ID: | 98403 |
Gene Name: | zinc finger protein 451 |
Gene Aliases: |
4930515K21Rik, 4933435G09Rik, AI596398, COASTER, Kiaa0576-hp, Znf451, mKIAA1702 |
Location: |
Chr.1:33761541-33815452 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp451 | NM_001290699.1 | NP_001277628.1 | ||
| NM_133817.3 | 4 | 491 | NP_598578.1 | |
| AK030088.1 | 4 | 324 | BAC26778.1 | |
| BC062154.1 | AAH62154.1 |
More Information
Set Membership: |
|

Genomic Map