Assay Details
Target Gene Details
Entrez Gene ID: | 226409 |
Gene Name: | zinc finger, RAN-binding domain containing 3 |
Gene Aliases: |
4933425L19Rik, AH2, AI316834, C730006D09 |
Location: |
Chr.1:127954179-128103049 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zranb3 | NM_001285945.1 | NP_001272874.1 | ||
| NM_027678.3 | 1 | 58 | NP_081954.1 | |
| AK016921.1 | BAB30499.1 | |||
| AK042139.1 | BAC31181.1 | |||
| AK083641.1 | 1 | 37 | BAC38978.1 | |
| AK084943.1 | 1 | 42 | BAC39315.1 | |
| AK085488.1 | 1 | 51 | ||
| BC066035.1 | AAH66035.1 | |||
| BC117921.1 | ||||
| BC117922.2 | 1 | 588 |
More Information
Set Membership: |
|

Genomic Map