Assay Details
Target Gene Details
Entrez Gene ID: | 319651 |
Gene Name: | ubiquitin specific peptidase 37 |
Gene Aliases: |
4932415L06Rik, C330008N13Rik |
Location: |
Chr.1:74435510-74544286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp37 | NM_176972.4 | NP_795946.2 | ||
| AK030013.1 | BAC26734.1 | |||
| AK035640.1 | BAC29135.1 | |||
| BC139091.1 |
More Information
Set Membership: |
|

Genomic Map