Assay Details
Target Gene Details
Entrez Gene ID: | 319651 |
Gene Name: | ubiquitin specific peptidase 37 |
Gene Aliases: |
4932415L06Rik, C330008N13Rik |
Location: |
Chr.1:74435510-74544286 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp37 | NM_176972.4 | 7 | 898 | NP_795946.2 |
| AK030013.1 | 7 | 899 | BAC26734.1 | |
| AK035640.1 | 6 | 882 | BAC29135.1 | |
| AK173210.1 | 3 | 374 | BAD32488.1 | |
| BC139091.1 | 7 | 840 |
More Information
Set Membership: |
|

Genomic Map