Assay Details
Target Gene Details
Entrez Gene ID: | 226409 |
Gene Name: | zinc finger, RAN-binding domain containing 3 |
Gene Aliases: |
4933425L19Rik, AH2, AI316834, C730006D09 |
Location: |
Chr.1:127954179-128103049 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zranb3 | NM_001285945.1 | 9 | 1122 | NP_001272874.1 |
| NM_027678.3 | 9 | 1261 | NP_081954.1 | |
| AK016921.1 | 9 | 1123 | BAB30499.1 | |
| AK042139.1 | 9 | 1229 | BAC31181.1 | |
| AK050039.1 | 5 | 542 | BAC34042.1 | |
| AK083641.1 | 9 | 1240 | BAC38978.1 | |
| AK084943.1 | 9 | 1245 | BAC39315.1 | |
| AK085488.1 | 3 | 449 | ||
| BC066035.1 | 9 | 1198 | AAH66035.1 | |
| BC117921.1 | 9 | 1076 | ||
| BC117922.2 | 9 | 2268 |
More Information
Set Membership: |
|

Genomic Map