Assay Details
Target Gene Details
Entrez Gene ID: | 67883 |
Gene Name: | UDP-glucuronate decarboxylase 1 |
Gene Aliases: |
1600025I13Rik, AI451869, AI649125, AW550562 |
Location: |
Chr.1:43748459-43827821 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uxs1 | NM_026430.3 | NP_080706.1 | ||
| AF399958.1 | AAK85410.1 | |||
| AK005536.1 | ||||
| AK075806.1 | BAC35974.1 | |||
| AK076166.1 | ||||
| AK152376.1 | BAE31165.1 | |||
| AK177105.1 | ||||
| AK184776.1 | ||||
| AK186705.1 | ||||
| AK187327.1 | ||||
| BC037049.1 | AAH37049.1 |
More Information
Set Membership: |
|

Genomic Map