Assay Details
Target Gene Details
Entrez Gene ID: | 98685 |
Gene Name: | tRNA methyltransferase 1 like |
Gene Aliases: |
1190005F20Rik, AW226554, C1orf25, Trm1-like, Trm1l |
Location: |
Chr.1:151428547-151458183 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trmt1l | NM_026876.3 | 9 | 1381 | NP_081152.2 |
| AK146088.1 | 9 | 1382 | BAE26891.1 | |
| AK216989.1 | 1 | 38 | ||
| BC022682.1 | 4 | 417 | AAH22682.1 | |
| BC037478.1 | 5 | 561 | AAH37478.1 | |
| BC110425.1 | 9 | 1364 | ||
| BC132299.1 | 9 | 1239 |
More Information
Set Membership: |
|

Genomic Map