Assay Details
Target Gene Details
Entrez Gene ID: | 226861 |
Gene Name: | hedgehog acyltransferase |
Gene Aliases: |
2810432O22Rik, AI462858, AP-2CRE, Skn, Tg(TFAP2A-cre)1Will |
Location: |
Chr.1:192512828-192771412 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hhat | NM_144881.4 | 6 | 871 | NP_659130.2 |
| AK028230.1 | 6 | 674 | BAC25829.1 | |
| AK052669.1 | BAC35090.1 | |||
| AK134661.1 | 6 | 714 | ||
| AK172126.1 | 6 | 718 | BAE42837.1 |
More Information
Set Membership: |
|

Genomic Map