Assay Details
Target Gene Details
Entrez Gene ID: | 17082 |
Gene Name: | interleukin 1 receptor-like 1 |
Gene Aliases: |
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2 |
Location: |
Chr.1:40404864-40465418 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rl1 | NM_001025602.3 | 3 | 492 | NP_001020773.1 |
| NM_001294171.1 | 3 | 325 | NP_001281100.1 | |
| NM_010743.3 | 3 | 492 | NP_034873.2 | |
| AK033417.1 | 3 | 345 | BAC28277.1 | |
| AK145118.1 | 3 | 346 | BAE26245.1 | |
| BC051445.1 | 3 | 302 | AAH51445.1 | |
| BY228016.1 | 3 | 326 | ||
| D13695.1 | 2 | 184 | BAA02854.1 | |
| M24843.1 | 3 | 333 | AAA40160.1 | |
| Y07519.1 | 3 | 336 | CAA68812.1 |
More Information
Set Membership: |
|

Genomic Map