Assay Details
Target Gene Details
Entrez Gene ID: | 71449 |
Gene Name: | methyltransferase like 13 |
Gene Aliases: |
5630401D24Rik |
Location: |
Chr.1:162533671-162548345 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl13 | NM_144877.1 | 6 | 1720 | NP_659126.1 |
| AK084750.1 | 6 | 1415 | ||
| AK168959.1 | 6 | 1927 | BAE40765.1 | |
| BC014872.1 | 6 | 1720 | AAH14872.1 |
More Information
Set Membership: |
|

Genomic Map