Assay Details
Target Gene Details
Entrez Gene ID: | 170755 |
Gene Name: | serum/glucocorticoid regulated kinase 3 |
Gene Aliases: |
2510015P22Rik, A330005P07Rik, Cisk, fy, fz |
Location: |
Chr.1:9798130-9902570 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk3 | NM_001037759.1 | 2 | 174 | NP_001032848.1 |
| NM_133220.2 | 3 | 293 | NP_573483.1 | |
| NM_177547.3 | 2 | 186 | NP_808215.2 | |
| AF312007.1 | 3 | 235 | AAG34115.1 | |
| AK028722.1 | 3 | 238 | ||
| AK030314.1 | 2 | 175 | BAC26895.1 | |
| AK031133.1 | 3 | 274 | BAC27269.1 | |
| AK031328.1 | 2 | 115 | BAC27349.1 | |
| AK143836.1 | 3 | 281 | ||
| AK146561.1 | 2 | 129 | BAE27261.1 | |
| AK151181.1 | 3 | 238 | BAE30182.1 | |
| AK151793.1 | 2 | 166 | BAE30694.1 | |
| AK151867.1 | 3 | 270 | BAE30755.1 | |
| AK151996.1 | 3 | 219 | BAE30860.1 | |
| AK166414.1 | 2 | 116 | ||
| AK166531.1 | 2 | 136 | ||
| AK171120.1 | 2 | 162 | BAE42262.1 | |
| AK171186.1 | 2 | 162 | BAE42298.1 | |
| AK171678.1 | 2 | 145 | ||
| AK171959.1 | 2 | 223 | BAE42748.1 | |
| AK172323.1 | 2 | 136 | BAE42945.1 | |
| BB856423.1 | 2 | 210 |
More Information
Set Membership: |
|

Genomic Map