Assay Details
Target Gene Details
Entrez Gene ID: | 72080 |
Gene Name: | suppressor APC domain containing 2 |
Gene Aliases: |
2010317E24Rik, 6030458L21Rik, AL033337, ang |
Location: |
Chr.2:25372035-25378213 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sapcd2 | NM_001081085.2 | NP_001074554.1 | ||
| NM_001290407.1 | NP_001277336.1 | |||
| AB197927.1 | BAD86554.1 | |||
| AK008577.1 | BAB25756.1 | |||
| AK154143.1 | ||||
| AK163973.1 | BAE37561.1 | |||
| AK164615.1 | BAE37847.1 | |||
| AK166625.1 | BAE38903.1 | |||
| BC070477.1 | AAH70477.1 |

Genomic Map