Assay Details
Target Gene Details
Entrez Gene ID: | 74243 |
Gene Name: | SLX4 interacting protein |
Gene Aliases: |
2210009G21Rik, 2410004I22Rik, C85661, Slxip |
Location: |
Chr.2:136891218-137082176 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slx4ip | NM_001038641.1 | NP_001033730.1 | ||
| NM_028201.2 | NP_082477.1 | |||
| NM_028834.2 | NP_083110.1 | |||
| AK008687.1 | BAB25833.1 | |||
| AK010392.1 | BAB26906.1 | |||
| AK050487.1 | ||||
| AK136226.1 | BAE22883.1 | |||
| BC067037.1 | AAH67037.1 | |||
| BC071202.1 | AAH71202.1 |

Genomic Map