Assay Details
Target Gene Details
Entrez Gene ID: | 69487 |
Gene Name: | NADH dehydrogenase (ubiquinone) complex I, assembly factor 5 |
Gene Aliases: |
2310003L22Rik |
Location: |
Chr.2:140170631-140205252 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf5 | NM_027093.4 | NP_081369.2 | ||
| AK009123.1 | BAB26088.1 | |||
| AK187586.1 | ||||
| AK188392.1 | ||||
| BC005630.1 | AAH05630.1 | |||
| CD363717.1 |

Genomic Map