Assay Details
Target Gene Details
Entrez Gene ID: | 68870 |
Gene Name: | adenylate kinase 8 |
Gene Aliases: |
1190002A17Rik, AK 8 |
Location: |
Chr.2:28698226-28813165 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ak8 | NM_001033874.2 | NP_001029046.2 | ||
| AK004439.1 | ||||
| BC100481.1 | AAI00482.1 | |||
| BC109352.2 | AAI09353.1 |
More Information
Set Membership: |
|

Genomic Map