Assay Details
Target Gene Details
Entrez Gene ID: | 394436 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A1 |
Gene Aliases: |
Gnt1, UDPGT 1-1, UGT1A01, Udpgt-1a, UgtBr1 |
Location: |
Chr.1:88211959-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a1 | NM_201645.2 | 4 | 1190 | NP_964007.2 |
| AA674349.1 | ||||
| AI097692.1 | 3 | 167 | ||
| AY227194.1 | 4 | 1190 | AAP48593.1 | |
| BC093516.1 | 4 | 1152 | AAH93516.1 | |
| JX026002.1 | 3 | 1057 | ||
| L02333.1 | 4 | 1162 | ||
| S64760.1 | 4 | 1162 | AAB26033.2 |
Target Gene Details
Entrez Gene ID: | 394430 |
Gene Name: | UDP glycosyltransferase 1 family, polypeptide A10 |
Gene Aliases: |
A13, Ugt1a13 |
Location: |
Chr.1:88055411-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a10 | NM_201641.2 | 4 | 1203 | NP_964003.2 |
| AY227201.1 | 4 | 1162 | AAP48600.1 | |
| BC089569.1 | 4 | 1203 | AAH89569.1 |
Target Gene Details
Entrez Gene ID: | 22236 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A2 |
Gene Aliases: |
UDPGT 1-2, Ugt1 |
Location: |
Chr.1:88200611-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a2 | NM_013701.3 | 4 | 1167 | NP_038729.1 |
| AK002629.1 | 4 | 1168 | BAB22243.1 | |
| AY227195.1 | 4 | 1177 | AAP48594.1 | |
| BC138676.1 | 4 | 1166 | ||
| BC145969.1 | 4 | 1166 | ||
| D87866.1 | 4 | 1141 | BAA13482.1 |
Target Gene Details
Entrez Gene ID: | 394433 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A5 |
Gene Aliases: |
- |
Location: |
Chr.1:88166012-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a5 | NM_201643.2 | 4 | 1169 | NP_964005.2 |
| AB094481.1 | 4 | 1148 | BAD11305.1 | |
| AY227196.1 | 4 | 1169 | AAP48595.1 | |
| BC139093.1 | 4 | 1149 | ||
| BC139094.1 | 4 | 1149 |
Target Gene Details
Entrez Gene ID: | 394435 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A6B |
Gene Aliases: |
A9', Ugt1a9 |
Location: |
Chr.1:88103257-88218998 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a6b | NM_201410.3 | 5 | 1258 | NP_958812.3 |
| AY227198.1 | 5 | 1214 | AAP48597.1 | |
| BC026561.1 | 6 | 1238 | AAH26561.1 |
Target Gene Details
Entrez Gene ID: | 394432 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A7C |
Gene Aliases: |
A10', A13, Ugt1a10 |
Location: |
Chr.1:88095001-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a7c | NM_201642.4 | 4 | 1255 | NP_964004.1 |
| AK128918.1 | 4 | 1181 | BAC87656.1 | |
| AK144599.1 | 4 | 1147 | BAE25958.1 | |
| AK153157.1 | 4 | 1145 | BAE31767.1 | |
| AK154973.1 | 4 | 1178 | ||
| AK159496.1 | 4 | 1193 | ||
| AK162317.1 | 4 | 1179 | ||
| AK165465.1 | 4 | 1179 | BAE38202.1 | |
| AY227199.1 | 4 | 1159 | AAP48598.1 | |
| BC141205.1 | 4 | 1164 | ||
| BC141206.1 | 4 | 1164 |
Target Gene Details
Entrez Gene ID: | 394434 |
Gene Name: | UDP glucuronosyltransferase 1 family, polypeptide A9 |
Gene Aliases: |
UDPGT, UDPGT 1-9, UGT1-09, UGT1.9, UGTP4, Ugt1a12, mUGTBr/P |
Location: |
Chr.1:88070779-88220002 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ugt1a9 | NM_201644.2 | 4 | 1177 | NP_964006.2 |
| AY227200.1 | 4 | 1156 | AAP48599.1 | |
| BC138699.1 | 4 | 1156 | ||
| BC146021.1 | 4 | 1156 | ||
| L27122.1 | 4 | 1102 | AAA40524.1 |
More Information
Set Membership: |
|

Genomic Map