Assay Details
Target Gene Details
Entrez Gene ID: | 22596 |
Gene Name: | X-ray repair complementing defective repair in Chinese hamster cells 5 |
Gene Aliases: |
AI314015, Ku80, Ku86 |
Location: |
Chr.1:72307417-72394953 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xrcc5 | NM_009533.2 | 21 | 2396 | NP_033559.2 |
| AF166486.1 | AAD49720.1 | |||
| AK089944.1 | 21 | 2394 | ||
| AK165470.1 | 21 | 2364 | BAE38207.1 | |
| AK167312.1 | 21 | 2285 | BAE39415.1 | |
| AK168913.1 | 21 | 2285 | BAE40726.1 | |
| AK177313.1 | ||||
| AK214786.1 | ||||
| BC029218.1 | 21 | 2268 | AAH29218.1 | |
| BC051660.1 | 21 | 2336 | AAH51660.1 | |
| CJ220465.1 | 3 | 223 | ||
| X66323.1 | 21 | 2297 | CAA46999.1 |
More Information
Set Membership: |
|

Genomic Map