Assay Details
Target Gene Details
Entrez Gene ID: | 269275 |
Gene Name: | activin A receptor, type IC |
Gene Aliases: |
ACVRLK7, ALK7, Alk-7, C230097P10 |
Location: |
Chr.2:58267453-58357896 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acvr1c | NM_001033369.3 | NP_001028541.1 | ||
| NM_001111030.1 | NP_001104500.1 | |||
| AK049089.1 | BAE43339.1 | |||
| AK142396.1 | ||||
| BC028780.1 | AAH28780.1 | |||
| BC145224.1 |
Target Gene Details
Entrez Gene ID: | 100502932 |
Gene Name: | predicted gene 13544 |
Gene Aliases: |
OTTMUSG00000012672 |
Location: |
Chr.2:58276779-58286902 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm13544 | NR_040365.1 | |||
| AK144626.1 | ||||
| AK165961.1 |
More Information
Set Membership: |
|

Genomic Map