Assay Details
Target Gene Details
Entrez Gene ID: | 11538 |
Gene Name: | activity-dependent neuroprotective protein |
Gene Aliases: |
AA589558, mKIAA0784 |
Location: |
Chr.2:168180965-168207062 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adnp | NM_009628.2 | NP_033758.2 | ||
| AK129214.1 | BAC98024.1 | |||
| AK134786.1 | BAE22284.1 |

Genomic Map