Assay Details
Target Gene Details
Entrez Gene ID: | 56369 |
Gene Name: | APAF1 interacting protein |
Gene Aliases: |
APIP2, CGI-29, Mmrp19 |
Location: |
Chr.2:103073500-103092649 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apip | NM_019735.4 | NP_062709.3 | ||
| AB028863.1 | BAA78906.1 | |||
| AK077705.1 | BAC36968.1 | |||
| AK145450.1 | BAE26445.1 | |||
| BC028434.1 | AAH28434.1 |
More Information
Set Membership: |
|

Genomic Map