Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf2 | NM_001025093.2 | NP_001020264.1 | ||
| NM_001284369.1 | NP_001271298.1 | |||
| NM_001284370.1 | NP_001271299.1 | |||
| NM_001284371.1 | NP_001271300.1 | |||
| NM_001284372.1 | NP_001271301.1 | |||
| NM_001284373.1 | NP_001271302.1 | |||
| NM_001284374.1 | NP_001271303.1 | |||
| NM_001284376.1 | NP_001271305.1 | |||
| NM_009715.3 | NP_033845.1 | |||
| AK035438.1 | BAC29065.1 | |||
| AK051591.1 | BAC34684.1 | |||
| AK087424.1 | BAC39869.1 | |||
| AK139515.1 | ||||
| AK139844.1 | BAE24156.1 | |||
| AK140855.1 | BAE24498.1 | |||
| AK144977.1 | ||||
| AK145887.1 | BAE26726.1 | |||
| AK146839.1 | ||||
| BC079883.1 | AAH79883.1 | |||
| BC082596.1 | AAH82596.1 |
More Information
Set Membership: |
|

Genomic Map