Assay Details
Target Gene Details
Entrez Gene ID: | 11909 |
Gene Name: | activating transcription factor 2 |
Gene Aliases: |
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP |
Location: |
Chr.2:73816509-73892639 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
More Information
Set Membership: |
|

Genomic Map