Assay Details
Target Gene Details
Entrez Gene ID: | 11949 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 |
Gene Aliases: |
1700094F02Rik |
Location: |
Chr.2:10056030-10080510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5c1 | NM_001112738.1 | NP_001106209.1 | ||
| NM_020615.4 | NP_065640.2 | |||
| AK007063.1 | BAB24848.1 | |||
| AK088164.1 | BAC40184.1 | |||
| AK146434.1 | BAE27168.1 | |||
| AK150307.1 | BAE29456.1 | |||
| AK158675.1 | BAE34606.1 | |||
| AK187037.1 | ||||
| BC010700.1 | AAH10700.1 | |||
| D21073.1 | BAB17293.1 |
More Information
Set Membership: |
|

Genomic Map