Assay Details
Target Gene Details
Entrez Gene ID: | 12075 |
Gene Name: | beaded filament structural protein 1, in lens-CP94 |
Gene Aliases: |
CP95 |
Location: |
Chr.2:143826528-143915274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bfsp1 | NM_001291061.1 | NP_001277990.1 | ||
| NM_009751.2 | NP_033881.2 | |||
| AB003147.1 | BAA24139.1 | |||
| AK053766.1 | BAC35514.1 | |||
| AK143180.1 | BAE25294.1 | |||
| BC057580.1 | AAH57580.1 |

Genomic Map