Assay Details
Target Gene Details
Entrez Gene ID: | 229003 |
Gene Name: | helicase with zinc finger 2, transcriptional coactivator |
Gene Aliases: |
PDIP1, Pric285, mKIAA1769, mPDIP1 |
Location: |
Chr.2:181227615-181242058 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Helz2 |

Genomic Map