Assay Details
Target Gene Details
Entrez Gene ID: | 212670 |
Gene Name: | cation channel, sperm associated 2 |
Gene Aliases: |
- |
Location: |
Chr.2:121393024-121416224 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Catsper2 | NM_153075.3 | NP_694715.2 | ||
| AF411816.1 | AAL26489.1 | |||
| AK142380.1 | BAE25048.1 | |||
| BC132140.1 |
More Information
Set Membership: |
|

Genomic Map