Assay Details
Target Gene Details
Entrez Gene ID: | 12287 |
Gene Name: | calcium channel, voltage-dependent, N type, alpha 1B subunit |
Gene Aliases: |
AW050276, AW060892, AW822256, Cav2.2, Cchn1a, alpha(1B) |
Location: |
Chr.2:24603860-24763195 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1b | NM_001042528.2 | NP_001035993.1 | ||
| NM_007579.3 | NP_031605.2 | |||
| AF042317.1 | AAB97840.1 | |||
| AK134542.1 | BAE22176.1 | |||
| U04999.1 | AAB60437.1 |
More Information
Set Membership: |
|

Genomic Map