Assay Details
Target Gene Details
Entrez Gene ID: | 381409 |
Gene Name: | cadherin-like 26 |
Gene Aliases: |
Gm1010 |
Location: |
Chr.2:178430515-178487366 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh26 | NM_001291189.1 | 3 | 499 | NP_001278118.1 |
| NM_198656.2 | 3 | 499 | NP_941058.1 | |
| AK132328.1 | 3 | 484 | BAE21107.1 | |
| BC053928.1 | 3 | 407 | AAH53928.1 |
More Information
Set Membership: |
|

Genomic Map