Assay Details
Target Gene Details
Entrez Gene ID: | 227526 |
Gene Name: | cerebral dopamine neurotrophic factor |
Gene Aliases: |
9330140G23, Armetl1 |
Location: |
Chr.2:3512966-3526376 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdnf | NM_177647.4 | NP_808315.1 | ||
| AK034009.1 | BAC28545.1 | |||
| BC110560.2 | AAI10561.1 |

Genomic Map