Assay Details
Target Gene Details
Entrez Gene ID: | 84682 |
Gene Name: | cytochrome c oxidase subunit IV isoform 2 |
Gene Aliases: |
Cox4b, CoxIV-2 |
Location: |
Chr.2:152754173-152765037 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox4i2 | NM_053091.2 | NP_444321.1 | ||
| AF271382.1 | AAK49433.1 | |||
| BC049623.1 | AAH49623.1 |
More Information
Set Membership: |
|

Genomic Map